Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Bicarbonate
SCI_ED

Catalog No. S25533B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
3 kg
Packaging:
Lab Grade
Reagent Grade
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25533B 3 kg Reagent Grade
S25533 500 g Reagent Grade
S25533A 2.5 kg Lab Grade
S25533C 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S25533B
Add to Cart
Add to Cart

Designed for educational use. Ideal for classroom experiments and instructional activities.

  • Product is for educational use
  • Available in 500 g, 2.5 kg, and 3 kg

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Quantity 3 kg
Assay Percent Range 99% w/v
Linear Formula NaHCO3
Formula Weight 84.01
Packaging Reagent Grade
Chemical Name or Material Sodium Bicarbonate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.