Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium bicarbonate anhydrous, free-flowing, Redi-Dri™, ACS reagent | 144-55-8 | MFCD00003528 | 1kg
Catalog No. 501883677
Encompass
Sodium bicarbonate anhydrous, free-flowing, Redi-Dri™, ACS reagent | Purity: 99.7% | MW: 84.01 | 144-55-8 | MFCD00003528 | 1kg
Catalog No. 50-188-3677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7925191KG
Sodium bicarbonate anhydrous, free-flowing, Redi-Dri™, ACS reagent | Purity: 99.7% | MW: 84.01 | 144-55-8 | MFCD00003528 | 1kg