Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, Concentrate 0.1 M NaHCO3 in Water, Eluent, Concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111013115 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3115 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3115 Supplier MilliporeSigma Supelco Supplier No. 364861L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 'bicarbonate concentrate' is basically 0.1-molar solution of sodium bicarbonate (the saturation temperature of the 0.1-molar solution is 15°C). It is highly advantageous as it is temperature dependent and its subsequent dilution with water can be appropriately controlled.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003528
Synonym Sodium bicarbonate solution

Specifications

CAS 144-55-8
Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.1 M NaHCO 3 aq. soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.