Learn More
Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | 270°C |
| Color | White |
| pH | 8.3 |
| Physical Form | Powder/Solid |
| Quantity | 12 kg |
| Solubility Information | Pass Test |
| Formula Weight | 84.01 |
| Percent Purity | 99.0 to 101.0% (EP) |
| Grade | BP/EP/FCC/JP/USP |
| Identification | Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP) |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:2 Flammability:0 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.