Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606332 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-332 45 kg Poly Bottle
18-606-327 500 g Poly Bottle
18-606-328 6 x 500 g Poly Bottle
18-606-330 4 x 2.5 kg Poly Bottle
18-606-331 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-332 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S114845KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1148, 144-55-8, NaHCO3

Spectrum™ Chemical Sodium Bicarbonate, Powder, FCC is used as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 45 kg
Percent Purity 99 to 100.5%
Grade FCC
Loss on Drying 0.0025
Packaging Poly Bottle
Chemical Name or Material Sodium Bicarbonate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.