Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Solstice

Catalog No. 6001988 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-88 1 kg
60-019-89 5 kg
60-044-800 6 x 1 kg
60-044-799 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-019-88 Supplier Solstice Supplier No. 134331KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Physical Form Powder
Quantity 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
Formula Weight 84.01g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Chemical Name or Material Sodium Bicarbonate, Meets analytical specification of Ph. Eur.,BP, USP, FCC
EINECSNumber 205-633-8
RTECSNumber VZ0950000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.