Learn More
Crescent Chemical Co Inc Sodium Bicarbonate, 144-55-8, MFCD00003528, 1kg
SDP
Catalog No. 50247908
Encompass_Preferred
Molecular Formula CHO3.Na, Molecular Weight 84.006, Research Grade, Ph. Eur., USP, Tested for use in tissue culture. Buffering substance. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-247-908 Supplier Crescent Chemical Co Inc Supplier No. SE30180.02
Molecular Formula CHO3.Na, Molecular Weight 84.006, Research Grade, Ph. Eur., USP, Tested for use in tissue culture. Buffering substance. Storage Conditions: +15 °C TO +30 °C