Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate/Sodium Carbonate Concentrate, IC eluent concentrate (20 x) for Metrosep A Supp 5, MilliporeSigma™ Supelco™

Catalog No. 111016047 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6047 1 L
11-100-4465 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6047 Supplier MilliporeSigma Supelco Supplier No. 624141L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Na2CO3 64 mM and NaHCO3 20 mM in Water

Prepared with sodium carbonate, sodium bicarbonate and high purity water (18.2 MOmega, 0.2 μm filtered)
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.