Learn More
Description
Sodium Bicarbonate USP is a white, crystalline powder. It is stable in dry air, but slowly decomposes in moist air.
Alternative name(s): Sodium hydrogen carbonate, baking soda
Specifications
Specifications
| Chemical Name or Material | Sodium Bicarbonate USP |
| Packaging | Jar, 600mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 144-55-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 84.0059 |
| Quantity | 500 g |
| Percent Purity | 99 to 100.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
