Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Biphenyl Complex

Catalog No. 2771341PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-713-41PAK 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 27-713-41PAK Supplier Sigma Aldrich Supplier No. 2771341PAK
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium biphenyl reagent is used to cleave the covalent fluorine bonds in organic compounds and biological samples. The liberated fluoride ions are determined spectrophotometrically or with the fluoride ion electrode.

Specifications

CAS 207121-48-0
Melting Point 163°C
Molecular Formula C3H7NO4S -+ H2O
Linear Formula HO2SCH2CH(NH2)CO2H-+H2O
MDL Number MFCD00150732
Quantity 1 g
Synonym Biphenyl sodium complex solution; Sodium biphenyl complex
Molecular Weight (g/mol) 171.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.