Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Bis(Trimethylsilyl)Amide Solution

Catalog No. 503081488 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1488 4 x 100 mL
24-558-525ML 4 x 25 mL
24-558-5100ML 100 mL
24-558-5800ML 800 mL
24-558-518L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 50-308-1488 Supplier Sigma Aldrich Supplier No. 2455854X100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium bis(trimethylsilyl)amide solution (NaHMDS) is widely used as a strong base in organic synthesis for deprotonation reactions and base-catalyzed reactions. It is also involved in the generation of enolates, Wittig reagents and carbenes.NaHMDS can be used as a base: In the synthesis of N-boc-1-amino-1,3-diene derivatives from N-boc-1-amino-4-methoxy-2-alkenes via 1,4-elimination reactions. To deprotonate alkyl or aryl ketones to generate enolate derivatives by hydroxylation reaction. In the enantioselective synthesis of 1,2-amino alcohols by the reaction between various nitroalkanes and aldehydes via nitroaldol reaction promoted by bimetallic heterogeneous catalyst.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1070-89-9
Density 0.904 g/mL at 25°C
Molecular Formula C6H18NNaSi2
Linear Formula [(CH3)3Si]2NNa
MDL Number MFCD00009835
Quantity 4 x 100 mL
Synonym Hexamethyldisilazane sodium salt solution
Molecular Weight (g/mol) 183.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.