Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium bismuthate
SCI_ED

Catalog No. s25534 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25534 10 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25534
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.967
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Quantity 10 g
Linear Formula NaBiO3
Formula Weight 279.97
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Sodium Bismuthate (V)
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.