Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfate, Anhydrous, Reagents

Catalog No. 01337056
Change view
Click to view available options
Quantity:
100 g
500 g
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-056 500 g
01-337-058 100 g
01-337-057 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-056 Supplier Reagents Holdings Llc Supplier No. C2309500500A5
 Restricted Item
Add to Cart
Add to Cart

Sodium Bisulfate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym Sodium hydrogen sulfate
IUPAC Name sodium hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.4394
Chemical Name or Material Sodium Bisulfate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.