Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfate AR Crystal, Macron Fine Chemicals™

Catalog No. MK7432500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7432-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-7432-500 Supplier Avantor Supplier No. 743212
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10034-88-5
Physical Form Crystalline
Quantity 500 g
Molecular Formula H3NaO5S
MDL Number MFCD00149210
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILES O.[Na+].OS([O-])(=O)=O
IUPAC Name sodium hydrate hydrogen sulfate
Molecular Weight (g/mol) 138.07
PubChem CID 23673662
Formula Weight 138.08
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Sodium Bisulfate, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.