Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bisulfate, Solstice

Catalog No. 6002848 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
600-28-48 250 g
600-28-49 5 kg
600-28-47 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 600-28-48 Supplier Solstice Supplier No. 71656250G
Add to Cart
Add to Cart

Purum, anhydrous, ∼95.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
IUPAC Name sodium;hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Quantity 250 g
Linear Formula NaHSO4
UN Number NONH for all modes of transport
Formula Weight 120.06g/mol
Percent Purity 93 to 102% (T), 95%
Chemical Name or Material Sodium bisulfate
Hazard Statement P280-P305 + P351 + P338
RTECSNumber VZ1860000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.