Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bisulfate monohydrate, 99%, for analysis

Catalog No. AC125005000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125005000 500 g Glass bottle
AC125000025 2.5 kg Plastic Bottle
AC125000100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125005000 Supplier Thermo Scientific Chemicals Supplier No. 125005000
  • $219.00
    $181.65 / Each of 1

    Save $37.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10034-88-5 ∣ H3NaO5S ∣ 138.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58.0°C
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 99%
Linear Formula NaHSO4·H2O
Merck Index 15,8722
Solubility Information Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4
IUPAC Name sodium;hydrogen sulfate;hydrate
Formula Weight 138.08
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Sodium bisulfate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.