Learn More
Electron Microscopy Sciences Sodium Bisulfite Solution 0.005m Aqueous 500 ML
SDP
Catalog No. 5031921
Encompass_Preferred
Used in Lillie's Method for Nucleic Acids (1965) for nuclear chromatin, and chromatin of plasmodia, etc. See references: Lillie, R.D., Histopathologic Technic and Practical Histochemistry, 3rd ed, McGraw-Hill, NY, pp 149-150, c. 1965 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd ed. McGraw-Hill, NY, p. 131, c. 1968.
Catalog No. 50-319-21 Supplier Electron Microscopy Sciences Supplier No. 2677403
Add to Cart
Add to Cart
Used in Lillie's Method for Nucleic Acids (1965) for nuclear chromatin, and chromatin of plasmodia, etc. See references: Lillie, R.D., Histopathologic Technic and Practical Histochemistry, 3rd ed, McGraw-Hill, NY, pp 149-150, c. 1965 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd ed. McGraw-Hill, NY, p. 131, c. 1968.