Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium bisulfite ACS reagent | 7631-90-5 | 500G
Catalog No. NC2183893
Encompass
Sodium bisulfite ACS reagent | Purity: >=58.5% (SO2) | Mol Wt: 104.06 | 7631-90-5 | 500G
Catalog No. NC2183893 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 243973500G
Add to Cart
Add to Cart
Sodium bisulfite ACS reagent | Purity: >=58.5% (SO2) | Mol Wt: 104.06 | 7631-90-5 | 500G