Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfite FCC, Macron Fine Chemicals™

Catalog No. MK7444212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7444-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7444-212 Supplier Avantor Supplier No. 744406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7631-90-5
Quantity 2.5 kg
Molecular Formula NaO3S
MDL Number MFCD00003530
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
SMILES [Na+].[O-]S([O-])=O
IUPAC Name sodium sulfite
Molecular Weight (g/mol) 103.05
PubChem CID 23665763
ChEBI CHEBI:26709
Formula Weight 104.06
Percent Purity 100%
Grade FCC
Packaging Poly
Chemical Name or Material Sodium Bisulfite, Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.