Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfite, Granular, FCC, 58.5-67.4%, Spectrum™ Chemical
SDP

Catalog No. 18606343 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-343 12 kg Poly Bottle
18-606-341 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-343 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S117212KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1172, 7631-90-5, NaHSO3

Spectrum™ Chemical Sodium Bisulfite, Granular, FCC is used as an antioxidant to preserve the aromas and flavors. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 12 kg
Percent Purity 58.5 to 67.4%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Bisulfite, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.