Learn More
Electron Microscopy Sciences Sodium Bisulfite Solution 1 LT
SDP
Catalog No. 5032080
Encompass_Preferred
Sodium Bisulfite solution is a prepared, ready-to-use, high quality solution that saves time and effort and produces consistent uniform results. It is a 1% aqueous stock solution for fungal staining.
Catalog No. 50-320-80 Supplier Electron Microscopy Sciences Supplier No. 2608406
Add to Cart
Add to Cart
Sodium Bisulfite solution is a prepared, ready-to-use, high quality solution that saves time and effort and produces consistent uniform results. It is a 1% aqueous stock solution for fungal staining.