Learn More
Electron Microscopy Sciences Sodium Bisulfite Solution 500 ML
SDP
Catalog No. 5032081
Encompass_Preferred
Sodium Bisulfite solution is a prepared, ready-to-use, high quality solution that saves time and effort and produces consistent uniform results. It is a 1% aqueous stock solution for fungal staining.
Catalog No. 50-320-81 Supplier Electron Microscopy Sciences Supplier No. 2608405
Sodium Bisulfite solution is a prepared, ready-to-use, high quality solution that saves time and effort and produces consistent uniform results. It is a 1% aqueous stock solution for fungal staining.