Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bitartrate, Monohydrate, BiotechGrade, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18606345 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-345 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-345 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1177500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1177, 6131-98-2, NaHC4H4O6·H2O

Spectrum™ Chemical Sodium Bitartrate, Monohydrate, BiotechGrade , also known as monosodium tartrate, is a sodium acid salt of tartaric acid and used as an acidity regulator as well as a test for ammonium cation. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 6131-98-2
Molecular Formula C4H7NaO7
Molecular Weight (g/mol) 190.08
InChI Key LLVQEXSQFBTIRD-UHFFFAOYNA-M
IUPAC Name sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
SMILES O.[Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

pH 2.5 to 4.0
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Percent Purity 99.5%
Grade Biotech
Packaging Poly Bottle
Chemical Name or Material Sodium Acetate, Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.