Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate, 10-Hydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003998 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-998 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-998 Supplier Avantor Supplier No. 357005
Add to Cart
Add to Cart
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 1303-96-4
Physical Form Powder
Quantity 2.5 kg
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Sodium Borate, 10-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.