Learn More
Growcells SODIUM BORATE 200MM 500ML PK2
Catalog No. 501034815
Encompass_Preferred
Borate buffered saline; 200mM Sodium Borate. Prepared with 18.2megOhm water, 0.2 micron sterile filtered and autoclaved in final packaging. Recipe/L: 47g Boric Acid, 8g Sodium Hydroxide. pH 8.0-8.4
Catalog No. 50-103-4815 Supplier Growcells Supplier No. MRGL11002
Borate buffered saline; 200mM Sodium Borate. Prepared with 18.2megOhm water, 0.2 micron sterile filtered and autoclaved in final packaging. Recipe/L: 47g Boric Acid, 8g Sodium Hydroxide. pH 8.0-8.4