Learn More
Electron Microscopy Sciences Sodium Borate 5% Solution 500 ML
SDP
Catalog No. 5031852
Encompass_Preferred
Used in Rhodanine Method for Copper, Mineral and Pigment stain. See Sheehan and Hrapchack, Theory and Practice of Histotechnology. St. Louis, The Mosby Company 1980 p. 230.
Catalog No. 50-318-52 Supplier Electron Microscopy Sciences Supplier No. 2660203
Used in Rhodanine Method for Copper, Mineral and Pigment stain. See Sheehan and Hrapchack, Theory and Practice of Histotechnology. St. Louis, The Mosby Company 1980 p. 230.