Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate ACS AR Granular, Macron Fine Chemicals™

Catalog No. MK7457212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7457-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7457-212 Supplier Avantor Supplier No. 745706
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1303-96-4
Physical Form Granules
Quantity 2.5 kg
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
Grade ACS
Packaging Poly
Chemical Name or Material Sodium Borate, 10-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.