Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Borate, Decahydrate
SCI_ED

Catalog No. s25537b Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25537B 500 g Lab Grade
S25537A 500 g Reagent Grade
S25537 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25537B
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1303-96-4
Synonym Borax, Sodium Tetraborate Decahydrate

Specifications

Color White
Quantity 500 g
Linear Formula Na2B4O7·10H2O
Formula Weight 381.42
Packaging Lab Grade
Density 1.73g/mL
Chemical Name or Material Sodium Borate, Decahydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.