Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate Decahydrate, 99.5-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19143605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19143605 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19143605 Supplier MP Biomedicals, Inc Supplier No. 0219143605
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1303-96-4
Molecular Formula Na2B4O7 · 10H2O

Specifications

Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.5-102.0%
Formula Weight 381.37
Grade ACS Reagent
Chemical Name or Material Sodium Borate Decahydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.