Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate, Decahydrate, 99.5 to 105.0%, ACS, MilliporeSigma™

Catalog No. MSX03555 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX03555 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX03555 Supplier MilliporeSigma Supplier No. SX03555
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS reagent grade sodium borate, decahydrate for analytical testing and laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula Na2B4O7·10H2O
Molecular Weight (g/mol) 381.37
Synonym Sodium Tetraborate, Dodecahydrate

Specifications

Color White
Physical Form Crystals or Granules
Quantity 12 kg
Assay Percent Range 99.5 to 105.0%
Formula Weight 381.37
Percent Purity 98%
Grade ACS
Chemical Name or Material Sodium Borate, Decahydrate, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.