Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate Decahydrate ACS MP Biomedicals

Catalog No. ICN19143601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19143601 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19143601 Supplier MP Biomedicals, Inc Supplier No. 0219143601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Synonym Sodium Tetraborate, Borax, Fused

Specifications

Quantity 1 kg
Density 2.367 g/mL
Formula Weight 381.37
Grade ACS
Chemical Name or Material Sodium Borate, Decahydrate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.