Learn More
Description
The term borax is used for a number of closely related minerals or chemical compounds that differ in their crystal water content, but usually refers to the decahydrate. Sodium borate is used for many applications:
- To make buffers, e.g. for gel electrophoresis of DNA, such as TBE (Tris/Borate/EDTA) or SB (Sodium borate) buffer, or as BBS (borate buffered saline) in coating procedures
- As a micronutrient fertilizer to correct boron-deficient soils
- To clean the brain cavity of a skull for mounting
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 99.5–105.0% |
| Solubility Information | 1g dissolves in 16M water, 0.6mL boiling water, ∼/mL glyceral; insoluble in alcohol. |
| Formula Weight | 381.37g/mol |
| Grade | Not Available |
| Chemical Name or Material | Sodium Borate Decahydrate |
Safety and Handling
| Recommended Storage | Store at Room Temperature(15–30 °C). |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.