Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate, Decahydrate, NF, 99-105%, Spectrum™ Chemical
SDP

Catalog No. 18606364 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-364 50 kg Poly Bottle
18-606-362 2.5 kg Poly Bottle
18-606-363 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-364 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S118350KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1183, 1303-96-4, Na2B4O7·10H2O

Spectrum™ Chemical Sodium Borate, Decahydrate, NF is an anti-fungal agent in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4H20Na2O17
Molecular Weight (g/mol) 381.36
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 99 to 105%
Grade NF
Packaging Poly Bottle
Chemical Name or Material Sodium Borate, Decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.