Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate Decahydrate, Purified (Borax; Sodium Tetraborate Decahydrate), Reagents

Catalog No. 01337051
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-051 500 g
01-337-052 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-051 Supplier Reagents Holdings Llc Supplier No. C2310500500A5
Add to Cart
Add to Cart

Sodium Borate Decahydrate, Purified (Borax; Sodium Tetraborate Decahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4H20Na2O17
Molecular Weight (g/mol) 381.36
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
Synonym Sodium Tetraborate, Borax
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

Specifications

Boiling Point 320°C
Color White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.73
Chemical Name or Material Sodium Borate Decahydrate, Purified (Borax, Sodium Tetraborate Decahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.