Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate Decahydrate, Technical, Spectrum™ Chemical
SDP

Catalog No. 18606354 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-354 50 kg Poly Bottle
18-606-351 500 g Poly Bottle
18-606-352 2.5 kg Poly Bottle
18-606-353 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-354 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S118650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1186, 1303-96-4, Na2B4O7·10H2O

Spectrum™ Chemical Sodium Borate Decahydrate, Technical is also known simply as Borax and is a mineral that occurs naturally in deposits of the mineral Evaporite. Its uses vary from household products, such as hand soap and water softening agents, to use as a flux in welding and can also be found in small scale gold mining operations as a substitute to mercury.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4H20Na2O17
Molecular Weight (g/mol) 381.36
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 50 kg
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Sodium Borate Decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.