Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borohydride, 97+%

Catalog No. p-4407862
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3578822 100 g
AA3578836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3578822 Supplier Thermo Scientific Chemicals Supplier No. 03578822
Only null left
Add to Cart

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases.Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in the ammonia borane dehydrogenation. Sodium borohydride is used as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces the aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydridoborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Granules
Quantity 100 g
Assay Percent Range >97+%
Solubility Information Soluble in water,ammonia and amines. Insoluble in ether.
Formula Weight 37.83
Sensitivity Moisture sensitive
Appearance White
Density 1.074 g/mL
Chemical Name or Material Sodium borohydride
Show More Show Less
Hazard Category H260-H301-H314-H360FD
Precautionary Statement P201-P202-P223-P231+P232-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
EINECSNumber 241-004-4
RTECSNumber ED3325000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.