Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals

Catalog No. ICN10289450 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10289450 50 g
ICN10289425 25 g
ICN10289480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10289450 Supplier MP Biomedicals, Inc Supplier No. 0210289450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Excellent reducing agent

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Spécifications

Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98-99%
Formula Weight 37.8
Chemical Name or Material Sodium Borohydride
missing translation for 'storageNote' Hygroscopic
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.