Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium borohydride, 98%, pellets (1-2 cm)
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | 360.0°C |
| Color | White |
| Physical Form | Pellets |
| Quantity | 2.5 kg |
| Assay Percent Range | 97.5% min. (NaBH4) |
| Linear Formula | NaBH4 |
| Fieser | 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189 |
| Merck Index | 15, 8727 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in ammonia- reacts with acids,soluble in amines,pyridine and dimethylether of,diethylene glycol,insol.in other ethers,hydrocarb.,alkyl chlorides,reacts with oxidizing agents |
| Formula Weight | 37.83 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts viol |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water,because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comf |
| EINECSNumber | 241-004-4 |
| RTECSNumber | ED3325000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction