Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, 98+% (Sodium tetrahydridoborate), Reagents

Catalog No. 01337050
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-050 100 g
01-337-049 10 g
01-337-048 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-050 Supplier Reagents Holdings Llc Supplier No. C2310550100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Borohydride, 98+% (Sodium tetrahydridoborate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium Tetrahydroborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point 400°C
Melting Point 400°C
Color White to Gray
Physical Form Solid
Quantity 100 g
Percent Purity ≥98%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.074
Chemical Name or Material Sodium Borohydride, 98+% (Sodium tetrahydridoborate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.