Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, Crystalline Powder, Reagent, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18606365 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
40 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-365 25 g Amber Glass Bottle
18-606-366 100 g Amber Glass Bottle
18-606-367 500 g Amber Glass Bottle
18-606-368 2.5 kg Amber Glass Bottle
18-606-370 40 kg Amber Glass Bottle
18-606-369 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-365 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S118725GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1187, 16940-66-2, NaBH4

Spectrum™ Chemical Sodium Borohydride, Crystalline Powder, Reagent, ACS is an inorganic compound that is white in color and usually found as a powder. It is a reducing agent with versatility and is used as a synthetic reagent in the laboratory. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Borohydride, Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.