Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Borohydride Solution

Catalog No. 45290425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-290-425ML 25 mL
45-290-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 45-290-425ML Supplier Sigma Aldrich Supplier No. 45290425ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alkaline sodium borohydride solution can be used as an efficient reagent for the generation of hydrogen via hydrolysis in the presence of various catalysts such as +a-Al2O3 supported cobalt catalysts, carbon supported cobalt catalysts, graphene oxide particles supported with cobalt nanoparticles and Ni45Au45Co10 trimetallic nanoparticles.Product of Ascensus

Specifications

CAS 16940-66-2
Quantity 25 mL
Molecular Formula H4BNa
Linear Formula NaBH4
MDL Number MFCD00003518
Molecular Weight (g/mol) 37.83 g/mol
Density 1.375 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.