Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromate, ≥99.5% (RT), MilliporeSigma™ Supelco™

Catalog No. 111019081 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9081 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9081 Supplier MilliporeSigma Supelco Supplier No. 71325250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium bromate is an organic compound which is sodium salt of bromic acid. This alkali metal bromate is mostly used as oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003476

Specifications

pH 5.0 to 9.0 (25°C, 50 mg/mL aq. soln.)
CAS 7789-38-0
Quantity 250 g
Linear Formula NaBrO3
Formula Weight 150.89
Percent Purity ≥99.5% (RT)
Grade Puriss. p.a.
Density 3.339 g/mL (at 25°C)
RTECSNumber EF8750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.