Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bromide AR Granular, Macron Fine Chemicals™

Catalog No. MK0535212 Shop All Avantor Products
Change view
Click to view available options
Poly; 4 x 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-0535-212 Poly; 4 x 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-0535-212 Supplier Avantor Supplier No. 053506

Specifications

CAS 7647-15-6
Physical Form Granules
Quantity 4 x 2.5 kg
Molecular Formula BrNa
MDL Number MFCD00003475
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES [Na+].[Br-]
IUPAC Name sodium bromide
Molecular Weight (g/mol) 102.89
PubChem CID 253881
ChEBI CHEBI:63004
Formula Weight 102.89
Packaging Poly
Chemical Name or Material Sodium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.