Learn More
Medchemexpress LLC Sodium butane-1-sulfonate | 2386-54-1 | MFCD00007540 | 98.0% | 160.17 g·mol⁻¹ | C4H9NaO3S | 100 MG
SDP
Catalog No. 5000301509
Encompass_Preferred
Sodium butane-1-sulfonate is the sodium salt of 1-butanesulfonic acid used as a reagent in chromatographic and analytical workflows. It is water-soluble and commonly employed as a mobile phase additive to improve separation of arsenic and selenium species in HPLC and related methods. Typical specifications include a molecular weight of 160.17 g·mol⁻¹ and high purity suitable for analytical applications.
  • Water-soluble sodium salt suitable for aqueous mobile phases.
  • Facilitates chromatographic separation of arsenic and selenium compounds.
  • High purity for analytical and HPLC applications.
  • Available in small-scale lab pack sizes for research use.
  • Stable solid form with molecular weight 160.17 g·mol⁻¹.
Catalog No. 50-003-01509 Supplier Medchemexpress LLC Supplier No. HYW017224100MG
Sodium butane-1-sulfonate is the sodium salt of 1-butanesulfonic acid used as a reagent in chromatographic and analytical workflows. It is water-soluble and commonly employed as a mobile phase additive to improve separation of arsenic and selenium species in HPLC and related methods. Typical specifications include a molecular weight of 160.17 g·mol⁻¹ and high purity suitable for analytical applications.
  • Water-soluble sodium salt suitable for aqueous mobile phases.
  • Facilitates chromatographic separation of arsenic and selenium compounds.
  • High purity for analytical and HPLC applications.
  • Available in small-scale lab pack sizes for research use.
  • Stable solid form with molecular weight 160.17 g·mol⁻¹.