Learn More
Medchemexpress LLC Sodium butane-1-sulfonate | 2386-54-1 | MFCD00007540 | 98.0% | 160.17 g/mol | C4H9NaO3S | 500 MG
SDP
Catalog No. 5000300568
Encompass_Preferred
Sodium butane-1-sulfonate is the sodium salt of butane-1-sulfonic acid used as an ionic mobile-phase additive for chromatographic separation of arsenic, selenium, and related analytes. It appears as a white to off-white crystalline powder, supplied for research use, and is commonly used to improve ion-pairing and retention in aqueous chromatographic methods.
  • Used as ionic mobile-phase additive for chromatographic separations.
  • Improves separation and resolution of arsenic and selenium species.
  • High purity suitable for analytical applications (manufacturer lists ~98% assay).
  • White to off-white crystalline powder; stable and easy to weigh.
  • Suits ion-pairing and retention control in aqueous methods.
Catalog No. 50-003-00568 Supplier Medchemexpress LLC Supplier No. HYW017224500MG
Sodium butane-1-sulfonate is the sodium salt of butane-1-sulfonic acid used as an ionic mobile-phase additive for chromatographic separation of arsenic, selenium, and related analytes. It appears as a white to off-white crystalline powder, supplied for research use, and is commonly used to improve ion-pairing and retention in aqueous chromatographic methods.
  • Used as ionic mobile-phase additive for chromatographic separations.
  • Improves separation and resolution of arsenic and selenium species.
  • High purity suitable for analytical applications (manufacturer lists ~98% assay).
  • White to off-white crystalline powder; stable and easy to weigh.
  • Suits ion-pairing and retention control in aqueous methods.