Learn More
Medchemexpress LLC Sodium butyrate | 156-54-7 | 99.8% | 500 MG
SDP
Catalog No. 5000407179
Encompass_Preferred
Sodium Butyrate is an inhibitor of HDAC, exhibiting anti-tumor activity. It induces morphological changes, inhibits cell proliferation, and impairs cell viability in nasopharyngeal carcinoma (NPC) cells. It is for research use only.
  • Inhibits HDAC activity.
  • Possesses anti-tumor activity.
  • Induces morphological changes and inhibits proliferation in NPC cells.
  • Cytotoxic to NPC cells (5-8F and 6-10B cell lines).
  • Promotes apoptosis in nasopharyngeal carcinoma cells.
  • Reduces cell viability and key protein levels (p-mTOR, PCNA).
  • Induces cell cycle arrest in the G0/G1 phase.
Catalog No. 50-004-07179 Supplier Medchemexpress LLC Supplier No. HYB0350A500MG
Sodium Butyrate is an inhibitor of HDAC, exhibiting anti-tumor activity. It induces morphological changes, inhibits cell proliferation, and impairs cell viability in nasopharyngeal carcinoma (NPC) cells. It is for research use only.
  • Inhibits HDAC activity.
  • Possesses anti-tumor activity.
  • Induces morphological changes and inhibits proliferation in NPC cells.
  • Cytotoxic to NPC cells (5-8F and 6-10B cell lines).
  • Promotes apoptosis in nasopharyngeal carcinoma cells.
  • Reduces cell viability and key protein levels (p-mTOR, PCNA).
  • Induces cell cycle arrest in the G0/G1 phase.