Learn More
Medchemexpress LLC Sodium butyrate | 156-54-7 | MFCD00002816 | 99.8% | 1 ML
SDP
Catalog No. 5000407142
Encompass_Preferred
Sodium Butyrate (sodium butanoate) is an inhibitor of Histone Deacetylase (HDAC) with established anti-tumor activity. This human endogenous and microbial metabolite induces morphological changes, inhibits cell proliferation, and impairs viability in nasopharyngeal carcinoma (NPC) cells. It is also known to induce NPC cell apoptosis by activating the mitochondrial apoptotic axis and influences cell cycle arrest.
  • Inhibits Histone Deacetylase (HDAC)
  • Exhibits anti-tumor activity
  • Induces morphological changes and inhibits cell proliferation in NPC cells
  • Promotes apoptosis in nasopharyngeal carcinoma cells
  • Decreases cell viability and affects p-mTOR and PCNA protein levels
  • Induces cell cycle arrest in G0/G1 phase
  • Provides neuroprotection and influences microglial cell polarization
  • Reverses SIRT2-related age phenotypes
Catalog No. 50-004-07142 Supplier Medchemexpress LLC Supplier No. HYB0350A10MM/1ML
Sodium Butyrate (sodium butanoate) is an inhibitor of Histone Deacetylase (HDAC) with established anti-tumor activity. This human endogenous and microbial metabolite induces morphological changes, inhibits cell proliferation, and impairs viability in nasopharyngeal carcinoma (NPC) cells. It is also known to induce NPC cell apoptosis by activating the mitochondrial apoptotic axis and influences cell cycle arrest.
  • Inhibits Histone Deacetylase (HDAC)
  • Exhibits anti-tumor activity
  • Induces morphological changes and inhibits cell proliferation in NPC cells
  • Promotes apoptosis in nasopharyngeal carcinoma cells
  • Decreases cell viability and affects p-mTOR and PCNA protein levels
  • Induces cell cycle arrest in G0/G1 phase
  • Provides neuroprotection and influences microglial cell polarization
  • Reverses SIRT2-related age phenotypes