Learn More
Selleck Chemical LLC Sodium butyrate 25mg 156-54-7
SDP
Supplier:  Selleck Chemical LLC S199925MG
Encompass_Preferred
Sodium butyrate (NaB), sodium salt of butyric acid, is a histone deacetylase inhibitor and competitively binds to the zinc sites of class I and II histone deacetylases (HDACs). Sodium butyrate (NaB) inhibits cell cycle progression, promotes differentiation, and induces apoptosis and autophagy in several types of cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7986