Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium butyrate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111013251 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3251 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3251 Supplier MilliporeSigma Supelco Supplier No. 07596100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 156-54-7
Product Line TraceCERT
Linear Formula CH3CH2CH2COONa
MDL Number MFCD00002816
Melting Point 250°C to 253°C
Synonym Butyric acid sodium salt
Quantity 100 mg
Formula Weight 110.09
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.