Learn More
Electron Microscopy Sciences Sodium Cacodylate Buffer, 0.2M, pH Specify 225 ML
SDP
Catalog No. 50980231
Encompass_Preferred
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02). Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Specify pH at time of order.
Standard pH: 7.4
Catalog No. 50-980-231 Supplier Electron Microscopy Sciences Supplier No. 11650
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02). Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Specify pH at time of order.
Standard pH: 7.4